accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13071-79-9

IUPAC Name: O,O-diethyl {[(tert-butylsulfanyl)methyl]sulfanyl}phosphonothioate
Synonyms: terbufos contraven caswell no. 131A counter epa pesticide chemical code 105001
Molecular Weight (g/mol): 288.42
Molecular Formula: C9H21O2PS3
InChi Key: XLNZEKHULJKQBA-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCSC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results