accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFSFFF

CAS RN 130783-02-7

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]sulfanide
Synonyms: [3,5-bis(trifluoromethyl)phenyl]sulfanide
Molecular Weight (g/mol): 245.16
Molecular Formula: C8H3F6S
InChi Key: KCAQWPZIMLLEAF-UHFFFAOYSA-M
SMILES: FC(F)(F)C1=CC(=CC([S-])=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results