accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CrCrOOO

CAS RN 1308-38-9

IUPAC Name: oxo[(oxochromio)oxy]chromium
Synonyms: chromia chromium sesquioxide chromium(III) sesquioxide green chromium oxide dichromium trioxide chromium oxide greens chrom(III)-oxid chromium(III) oxide green oxide of chromium
Molecular Weight (g/mol): 151.99
Molecular Formula: Cr2O3
InChi Key: QDOXWKRWXJOMAK-UHFFFAOYSA-N
SMILES: O=[Cr]O[Cr]=O
Quantity 
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (6)
Physical Form 
  • (2)
  • (2)
  • (10)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (13)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (10)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)