accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COPOOCH3CH3H3C

CAS RN 13086-84-5

IUPAC Name: bis(tert-butoxy)(oxo)-λ⁵-phosphanylium
Synonyms: bis(tert-butoxy)(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 193.2
Molecular Formula: C8H18O3P
InChi Key: GEBLOQXLELCEEO-UHFFFAOYSA-N
SMILES: CC(C)(C)O[P+](=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)