accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1309-56-4

SMoS

CAS RN 1309-56-4

IUPAC Name: disulfanylidenemolybdenum
Synonyms: molybdenum disulfide MoS2 molybdenum disulphide molybdenum sulfide
Molecular Weight (g/mol): 160.07
Molecular Formula: MoS2
InChi Key: CWQXQMHSOZUFJS-UHFFFAOYSA-N
SMILES: S=[Mo]=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results