accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13096-96-3

ClHNN

CAS RN 13096-96-3

IUPAC Name: 4-chloro-1H-indazole
Synonyms: 4-chloro-1H-indazole
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: CQTGQYVQJOJQCM-UHFFFAOYSA-N
SMILES: ClC1=C2C=NNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results