accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131-58-8

IUPAC Name: (2-methylphenyl)(phenyl)methanone
Synonyms: (2-methylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: CKGKXGQVRVAKEA-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Certifications/Compliance 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results