accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1310-53-8

IUPAC Name: germanedione
Synonyms: germanium dioxide germania
Molecular Weight (g/mol): 104.63
Molecular Formula: GeO2
InChi Key: YBMRDBCBODYGJE-UHFFFAOYSA-N
SMILES: O=[Ge]=O
Molecular Weight (g/mol) 
  • (3)
  • (9)
Physical Form 
  • (9)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (2)
  • (3)
  • (9)
  • (9)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)