accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1310-65-2

LiOH

CAS RN 1310-65-2

IUPAC Name: lithium(1+) hydroxide
Synonyms: lithium hydroxide LiOH lithium hydroxide anhydrous lithium hydrate lithiumhydroxid
Molecular Weight (g/mol): 23.95
Molecular Formula: HLiO
InChi Key: WMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES: [Li+].[OH-]
Quantity 
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (3)
Physical Form 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)