accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13104-56-8

IUPAC Name: 4-(4-methoxyphenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms: 4-(4-methoxyphenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 339.4
Molecular Formula: C22H17N3O
InChi Key: QAEWAYWLMREGRA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results