accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131052-46-5

IUPAC Name: hydrogen 1-(1,3-thiazol-5-yl)methanamine chloride
Synonyms: hydrogen 1-(1,3-thiazol-5-yl)methanamine chloride
Molecular Weight (g/mol): 150.62
Molecular Formula: C4H7ClN2S
InChi Key: WZJFBMHEYMPACF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CN=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results