CAS RN 1312-81-8
Lanthanum(III) oxide, 99.9%
CAS: 1312-81-8 | La{2}O{3}
Lanthanum(III) oxide, 99.99%, (trace metal basis)
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) oxide, REacton™, 99.99% (REO)
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
Lanthanum(III) oxide, 99.9% (metals basis)
CAS: 1312-81-8 MDL Number: MFCD00011071
Lanthanum(III) oxide, REacton™, 99.9% (REO)
CAS: 1312-81-8 MDL Number: MFCD00011071 Synonym: Lanthanum trioxide,Lanthanum sesquioxide
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
CAS: 1312-81-8 Molecular Formula: La2O3 in 5% HNO3 MDL Number: MFCD00011066
Lanthanum(III) oxide, 99.9%, (trace metal basis)
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) oxide, REacton™, 99.999% (REO)
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071
Lanthanum(III) oxide, 91%
CAS: 1312-81-8 Molecular Formula: La2O3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011071
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade
CAS: 1312-81-8 Molecular Formula: La2O3 MDL Number: MFCD00011071