CAS RN 1313-13-9
Manganese(IV) oxide, 99.9% (metals basis)
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) Oxide, 99%, -10 mesh
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, 97%
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide activated, tech. 90%
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, 88%, electrolytically precipitated, active
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, 99.99+%, (trace metal basis)
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, activated, tech., Mn 58% min
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, 80-85%, powder
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Manganese(IV) oxide activated, Solstice
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
Manganese(IV) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Manganese(IV) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Manganese(Iv) Oxide, Extra Pure, SLR, Precipitated, Fisher Chemical™
CAS: 1313-13-9 Molecular Formula: MnO2 Molecular Weight (g/mol): 86.94 MDL Number: MFCD00003463 InChI Key: NUJOXMJBOLGQSY-UHFFFAOYSA-N Synonym: manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein PubChem CID: 14801 IUPAC Name: dioxomanganese SMILES: O=[Mn]=O
- 1
- 2