accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1313-49-1

NZnNZnZn

CAS RN 1313-49-1

IUPAC Name: zinc(2+) bis(zincioazanide)
Synonyms: zinc nitride
Molecular Weight (g/mol): 224.15
Molecular Formula: N2Zn3
InChi Key: AKJVMGQSGCSQBU-UHFFFAOYSA-N
SMILES: [Zn++].[N-]=[Zn].[N-]=[Zn]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results