accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaS

CAS RN 1313-82-2

IUPAC Name: disodium sulfanide
Synonyms: sodium sulfide
Molecular Weight (g/mol): 79.05
Molecular Formula: HNa2S
InChi Key: VDQVEACBQKUUSU-UHFFFAOYSA-M
SMILES: [Na+].[Na+].[SH-]
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)