accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONbOONbOO

CAS RN 1313-96-8

IUPAC Name: [(dioxoniobio)oxy]dioxoniobium
Synonyms: niobium pentoxide
Molecular Weight (g/mol): 265.81
Molecular Formula: Nb2O5
InChi Key: ZKATWMILCYLAPD-UHFFFAOYSA-N
SMILES: O=[Nb](=O)O[Nb](=O)=O
Physical Form 
  • (10)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (3)
  • (10)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)