accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1313-97-9

IUPAC Name: dineodymium(3+) trioxidandiide
Synonyms: neodymium(III) oxide
Molecular Weight (g/mol): 336.48
Molecular Formula: Nd2O3
InChi Key: PLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Nd+3].[Nd+3]
Physical Form 
  • (15)
  • (2)
Quantity 
  • (2)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
Grade 
  • (10)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (17)
  • (15)
  • (2)
  • (2)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (10)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)