CAS RN 1313-99-1
CAS RN 1313-99-1
Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3})
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, Puratronic™, 99.998% (metals basis)
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, green, Ni 78.5%
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, black, 76% Ni
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, 97%, green, reagent, powder
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, 99% (metals basis)
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]
Nickel(II) oxide, 99.995% (metals basis)
CAS: 1313-99-1 Molecular Formula: NiO Molecular Weight (g/mol): 74.69 MDL Number: MFCD00011145 InChI Key: GNRSAWUEBMWBQH-UHFFFAOYSA-N Synonym: nickel ii oxide,nickel monoxide,bunsenite,nickelous oxide,nickel oxide,mononickel oxide,nickel protoxide,black nickel oxide,nickel 2+ oxide PubChem CID: 14805 IUPAC Name: oxonickel SMILES: O=[Ni]