accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13138-33-5

IUPAC Name: (3-aminopropyl)phosphonic acid
Synonyms: 3-aminopropylphosphonic acid
Molecular Weight (g/mol): 139.09
Molecular Formula: C3H10NO3P
InChi Key: GSZQTIFGANBTNF-UHFFFAOYSA-N
SMILES: NCCCP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results