accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13139-12-3

H3CH3CCH3OOONOO

CAS RN 13139-12-3

IUPAC Name: tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate
Synonyms: tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Weight (g/mol): 215.21
Molecular Formula: C9H13NO5
InChi Key: VTGFSVGZCYYHLO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)