accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1314-08-5

OPd

CAS RN 1314-08-5

IUPAC Name: palladium(2+) oxidandiide
Synonyms: palladium(II) oxide
Molecular Weight (g/mol): 122.42
Molecular Formula: OPd
InChi Key: JQPTYAILLJKUCY-UHFFFAOYSA-N
SMILES: [O--].[Pd++]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)