accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OPbOPbOPbO

CAS RN 1314-41-6

IUPAC Name: bicyclo[3.1.1]triplumboxan-1-yl
Synonyms: Lead(II,IV) oxide
Molecular Weight (g/mol): 685.6
Molecular Formula: O4Pb3
InChi Key: XMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILES: O1[Pb]2O[Pb]1O[Pb]O2
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (4)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)