accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSbOOSbOO

CAS RN 1314-60-9

IUPAC Name: [(dioxostibanyl)oxy]stibanedione
Synonyms: antimony pentoxide
Molecular Weight (g/mol): 323.52
Molecular Formula: O5Sb2
InChi Key: LJCFOYOSGPHIOO-UHFFFAOYSA-N
SMILES: O=[Sb](=O)O[Sb](=O)=O
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)