accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OTaOTaOOO

CAS RN 1314-61-0

IUPAC Name: ditantalum(5+) pentaoxidandiide
Synonyms: ditantalum(5+) pentaoxidandiide
Molecular Weight (g/mol): 441.89
Molecular Formula: O5Ta2
InChi Key: BPUBBGLMJRNUCC-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Ta+5].[Ta+5]
Physical Form 
  • (12)
  • (2)
Quantity 
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
Grade 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (12)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)