accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1314-68-7

OReOOOReOOO

CAS RN 1314-68-7

IUPAC Name: trioxo[(trioxorhenio)oxy]rhenium
Synonyms: rhenium(VII) oxide
Molecular Weight (g/mol): 484.41
Molecular Formula: O7Re2
InChi Key: NBGOSNNAAHDRLK-UHFFFAOYSA-N
SMILES: O=[Re](=O)(=O)O[Re](=O)(=O)=O
Physical Form 
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results