accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1314-91-6

TePb

CAS RN 1314-91-6

IUPAC Name: plumbanetellone
Synonyms: lead telluride
Molecular Weight (g/mol): 334.8
Molecular Formula: PbTe
InChi Key: OCGWQDWYSQAFTO-UHFFFAOYSA-N
SMILES: [Te]=[Pb]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)