CAS RN 1317-36-8
Lead(II) oxide, Puratronic™, 99.999% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: oxolead SMILES: O=[Pb]
Lead(II) oxide, low silver
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) oxide, 99.9%
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: oxolead SMILES: O=[Pb]
Lead(II) oxide, 99.99% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) oxide, 99+%
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]
Lead(II) oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Solstice
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Lead(Ii) Oxide, ReagentPlus™, ≥99.9% trace metals basis, Honeywell™
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: oxolead SMILES: O=[Pb]
Lead(II) oxide, Fluka™
Puriss. p.a., ≥99.0% (KT), yellow
Lead Oxide, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]
Lead(II) oxide, ACS Reagent, ≥99.0%, Honeywell Fluka™
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
- 1
- 2