accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FeFeFeOOOO

CAS RN 1317-61-9

IUPAC Name: oxo[(oxoferrio)oxy]iron; oxoiron
Synonyms: Iron(II,III) oxide
Molecular Weight (g/mol): 231.53
Molecular Formula: Fe3O4
InChi Key: SZVJSHCCFOBDDC-UHFFFAOYSA-N
SMILES: O=[Fe].O=[Fe]O[Fe]=O
Physical Form 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)