CAS RN 13171-64-7
CAS RN 13171-64-7
IUPAC Name:
1,4-dimethyl (2S,3S)-2,3-dihydroxybutanedioate
Synonyms:
1,4-dimethyl (2S,3S)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol):
178.14
Molecular Formula:
C6H10O6
InChi Key:
PVRATXCXJDHJJN-IMJSIDKUSA-N
SMILES:
COC(=O)[C@@H](O)[C@H](O)C(=O)OC
1
–
1
of
1
results
1
Dimethyl D-tartrate, 99%
CAS: 13171-64-7 Molecular Formula: C6H10O6 Molecular Weight (g/mol): 178.14 MDL Number: MFCD00008445 InChI Key: PVRATXCXJDHJJN-IMJSIDKUSA-N Synonym: dimethyl d-tartrate,--dimethyl d-tartrate,dimethyl d---tartrate,dimethyl 2s,3s-2,3-dihydroxybutanedioate,butanedioic acid, 2,3-dihydroxy-, dimethyl ester, 2s,3s,d---tartaric acid dimethyl ester,dl-dimethyl tartrate,dl-dimethyl tartarate,d--dimethyl d-tartrate,meso-tartaric acid, dimethyl ester PubChem CID: 21142 IUPAC Name: dimethyl (2S,3S)-2,3-dihydroxybutanedioate SMILES: COC(=O)C(C(C(=O)OC)O)O