accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FClOO

CAS RN 131728-94-4

IUPAC Name: 8-(chloromethyl)-6-fluoro-2,4-dihydro-1,3-benzodioxine
Synonyms: 8-(chloromethyl)-6-fluoro-2,4-dihydro-1,3-benzodioxine
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: FMONGDHUPLQOCP-UHFFFAOYSA-N
SMILES: FC1=CC(CCl)=C2OCOCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results