accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131738-73-3

CH3H3COBr

CAS RN 131738-73-3

IUPAC Name: 1-bromo-3-(propan-2-yloxy)benzene
Synonyms: 1-bromo-3-isopropoxybenzene
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: JYWJZIQSPRFCDB-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results