accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131819-23-3

IUPAC Name: 4-propyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-propyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 338.46
Molecular Formula: C21H29F3
InChi Key: FEWMLRARKGRCCE-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results