CAS RN 1319-46-6
IUPAC Name:
tris(λ²-lead(2+)) dihydroxide dicarbonate
Synonyms:
white lead
Molecular Weight (g/mol):
775.6
Molecular Formula:
C2H2O8Pb3
InChi Key:
RYZCLUQMCYZBJQ-UHFFFAOYSA-H
SMILES:
[OH-].[OH-].[Pb++].[Pb++].[Pb++].[O-]C([O-])=O.[O-]C([O-])=O
1
–
5
of
5
results
1
Lead(II) Carbonate Basic, Pure, SLR, Precipitated, Fisher Chemical™
CAS: 1319-46-6 Molecular Formula: C2H2O8Pb3 MDL Number: 78155
2
Lead(II) carbonate, basic, ACS, Thermo Scientific Chemicals
CAS: 1319-46-6 MDL Number: MFCD00078155 Synonym: White lead; C.I. 77597
3
Lead(II) carbonate, basic, extra pure
CAS: 1319-46-6 Molecular Formula: C2H2O8Pb3 Molecular Weight (g/mol): 775.59 MDL Number: MFCD00078155
4
Lead(II) carbonate, basic, 99% (metals basis)
CAS: 1319-46-6 Molecular Formula: C2H2O8Pb3 MDL Number: MFCD00078155
5
Lead(Ii) Carbonate Basic, Solstice
CAS: 1319-46-6 MDL Number: MFCD00078155