accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNHFFF

CAS RN 131922-05-9

IUPAC Name: 2-(trifluoromethyl)piperazine
Synonyms: 2-(trifluoromethyl)piperazine
Molecular Weight (g/mol): 154.14
Molecular Formula: C5H9F3N2
InChi Key: YNEJOOCZWXGXDU-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CNCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results