accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131947-13-2

IUPAC Name: dichloro-1,2-thiazole-3-carboxylic acid
Synonyms: dichloro-1,2-thiazole-3-carboxylic acid
Molecular Weight (g/mol): 198.02
Molecular Formula: C4HCl2NO2S
InChi Key: ZFEHQZVNKOESSZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=NSC(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results