accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131986-28-2

IUPAC Name: 3-(4-chloro-1,2,5-thiadiazol-3-yl)pyridine
Synonyms: 3-(4-chloro-1,2,5-thiadiazol-3-yl)pyridine
Molecular Weight (g/mol): 197.64
Molecular Formula: C7H4ClN3S
InChi Key: CMPNWGQBNRHIQZ-UHFFFAOYSA-N
SMILES: ClC1=NSN=C1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results