accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 132-60-5

IUPAC Name: 2-phenylquinoline-4-carboxylic acid
Synonyms: tervalon aciphenochinoline aciphenochinolium artamin aciphenochinolinum cincain acifenokinolin cinchophen (substance) cinchophenum cincofeno
Molecular Weight (g/mol): 249.27
Molecular Formula: C16H11NO2
InChi Key: YTRMTPPVNRALON-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results