accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 132-64-9

IUPAC Name: 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: dibenzofuran 2,2'-biphenylylene oxide DBF 2,2'-biphenylene oxide dibenzol(b,d)furan diphenylene oxide dibenzo(b,d)furan
Molecular Weight (g/mol): 168.2
Molecular Formula: C12H8O
InChi Key: TXCDCPKCNAJMEE-UHFFFAOYSA-N
SMILES: O1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)