accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CNCH3ONN

CAS RN 132-69-4

IUPAC Name: hydrogen {3-[(1-benzyl-1H-indazol-3-yl)oxy]propyl}dimethylamine chloride
Synonyms: benzydamine
Molecular Weight (g/mol): 345.87
Molecular Formula: C19H24ClN3O
InChi Key: HNNIWKQLJSNAEQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCOC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)