accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSOHO

CAS RN 13205-48-6

IUPAC Name: 4-(methylsulfanyl)benzoic acid
Synonyms: P-methiobenzoic acid
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: KWHCPERWLHBLOT-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results