CAS RN 132119-11-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl (5-Benzoyl-1-methyl-1H-benzimidazol-2-yl)carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1-Methyl Mebendazole, TRC
CAS: 132119-11-0 Molecular Formula: C17 H15 N3 O3 Molecular Weight (g/mol): 309.32 Synonym: Methyl (5-Benzoyl-1-methyl-1H-benzimidazol-2-yl)carbamate,Mebendazole Imp. D (EP) IUPAC Name: methyl N-(5-benzoyl-1-methylbenzimidazol-2-yl)carbamate SMILES: COC(=O)Nc1nc2cc(ccc2n1C)C(=O)c3ccccc3