accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 132327-80-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(triphenylmethyl)carbamoyl]butanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(triphenylmethylcarbamoyl)butanoic acid
Molecular Weight (g/mol): 610.71
Molecular Formula: C39H34N2O5
InChi Key: WDGICUODAOGOMO-UHFFFAOYNA-N
SMILES: OC(=O)C(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)