accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FFFFN

CAS RN 1323966-11-5

IUPAC Name: 2-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 217.17
Molecular Formula: C10H7F4N
InChi Key: CBMBAVZSZLOOAE-UHFFFAOYSA-N
SMILES: CC1=CC(CC#N)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results