accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1323966-32-0

FOFFFN

CAS RN 1323966-32-0

IUPAC Name: 4-fluoro-2-(trifluoromethoxy)benzonitrile
Synonyms: 4-fluoro-2-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: HGQQVVYULDWWOU-UHFFFAOYSA-N
SMILES: FC1=CC(OC(F)(F)F)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results