accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1324063-28-6

CH3OOHFFFF(E/Z)

CAS RN 1324063-28-6

IUPAC Name: 3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 248.18
Molecular Formula: C11H8F4O2
InChi Key: BCBIVYJBNVUHLY-UHFFFAOYSA-N
SMILES: CC1=C(C=C(F)C(C=CC(O)=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results