accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOOHOHN(E/Z)

CAS RN 132465-10-2

IUPAC Name: 2-(thiophen-2-yl)ethyl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms: 2-(thiophen-2-yl)ethyl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 315.34
Molecular Formula: C16H13NO4S
InChi Key: KCGLTUBCAGZLHP-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)OCCC2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results