accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HgOOFFFOOFFF

CAS RN 13257-51-7

IUPAC Name: mercury(2+) ditrifluoroacetate
Synonyms: mercury(2+) ditrifluoroacetate
Molecular Weight (g/mol): 426.62
Molecular Formula: C4F6HgO4
InChi Key: WMHOESUUCMEQMS-UHFFFAOYSA-L
SMILES: [Hg++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)