accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2N

CAS RN 13258-63-4

IUPAC Name: 2-(pyridin-4-yl)ethan-1-amine
Synonyms: 4-pyridine ethaneamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: IDLHTECVNDEOIY-UHFFFAOYSA-N
SMILES: NCCC1=CC=NC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)