accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OAsOAsO

CAS RN 1327-53-3

IUPAC Name: diarsenic(3+) trioxidandiide
Synonyms: arsenic trioxide
Molecular Weight (g/mol): 197.84
Molecular Formula: As2O3
InChi Key: QTLQKAJBUDWPIB-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[As+3].[As+3]
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (4)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (11)
Grade 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (11)
  • (2)
  • (1)
  • (3)