accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 132741-81-2

CAS RN 132741-81-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results